Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(dimethylamino)phosphine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16248-0100 16248-0500 16248-1000
TS16248-0100EA 95 USD InStock
TS16248-0100 TS16248-0500 TS16248-1000
Tris(dimethylamino)phosphine 97%
Tris(dimethylamino)phosphine


Formula: C₆H₁₈N₃P
MW: 163.2 g/mol
Boiling Pt: 150 °C (760 mmHg)
Melting Pt: –44 °C
Density: 0.89 g/cm³
Flash Pt: 26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008301
CAS Number: 1608-26-0
EINECS: 216-534-4
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4670 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty