Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzene-1,3,5-tricarbonyl trichloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16437-0050 16437-1000 16437-5000
TS16437-0050EA 36 USD InStock
TS16437-0050 TS16437-1000 TS16437-5000
Benzene-1,3,5-tricarbonyl trichloride 98%
Benzene-1,3,5-tricarbonyl trichloride


Formula: C₉H₃Cl₃O₃
MW: 265.48 g/mol
Boiling Pt: 312
Melting Pt: 34.5…36 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000679
CAS Number: 4422-95-1
EINECS: 224-594-8
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance White to yellow Solid or liquid
Infrared spectrum Conforms
Melting point 30 to 35 °C
Titration Argentometric 97.5 to 102.5 % (ex Cl)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty