Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Furoyl chloride 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16538-0250 16538-1000
TS16538-0250EA 61 USD InStock
TS16538-0250 TS16538-1000
2-Furoyl chloride 98+%
2-Furoyl chloride


Formula: C₅H₃ClO₂
MW: 130.53 g/mol
Boiling Pt: 173…174 °C (760 mmHg)
Melting Pt: –2 °C
Density: 1.32 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00003228
CAS Number: 527-69-5
EINECS: 208-422-9
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5320 (20°C, 589 nm)
GC ≥98.0 %
Titration RCOCl ≥98.0 %
Impurity ≤2.0 % Free carboxylic acid (as furoic acid)
Free acid (HCl) ≤0.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty