Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lauroyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16903-0010 16903-0500 16903-2500
TS16903-0010EA 228 USD InStock
TS16903-0010 TS16903-0500 TS16903-2500
Lauroyl chloride 98%
Lauroyl chloride


Formula: C₁₂H₂₃ClO
MW: 218.77 g/mol
Boiling Pt: 267
Melting Pt: -12
Density: 0.92 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00000740
CAS Number: 112-16-3
EINECS: 203-941-7
UN: 3265
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤60 APHA

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty