Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexanoyl chloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16904-0050 16904-1000 16904-5000
TS16904-0050EA 25 USD InStock
TS16904-0050 TS16904-1000 TS16904-5000
Hexanoyl chloride 97%
Hexanoyl chloride


Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Boiling Pt: 152
Melting Pt: -87
Density: 0.95 g/cm³
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00000760
CAS Number: 142-61-0
EINECS: 205-549-1
UN: 2920
ADR: 8 II
Merck Index: 13,01769
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4270 (20°C, 589 nm)
GC ≥96.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty