Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isovaleryl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16928-0250 16928-1000 16928-5000
TS16928-0250EA 37 USD InStock
TS16928-0250 TS16928-1000 TS16928-5000
Isovaleryl chloride 98%
Isovaleryl chloride


Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 115
Density: 0.98 g/cm³
Flash Pt: 27 °C
MDL Number: MFCD00000738
CAS Number: 108-12-3
EINECS: 203-552-2
UN: 2920
ADR: 8 II
Merck Index: 13,05253
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty