Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-3-Methylcyclopentanone 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16937-0050 16937-0250
TS16937-0050EA 96 USD InStock
TS16937-0050 TS16937-0250
(±)-3-Methylcyclopentanone 99%
(±)-3-Methylcyclopentanone


Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 145
Density: 0.91 g/cm³
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00001416
CAS Number: 1757-42-2
EINECS: 217-148-9
UN: 1224
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty