Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Undec-10-enoyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17158-1000 17158-5000
TS17158-1000EA 109 USD InStock
TS17158-1000 TS17158-5000
Undec-10-enoyl chloride 98%
Undec-10-enoyl chloride


Formula: C₁₁H₁₉ClO
MW: 202.72 g/mol
Boiling Pt: 120
Density: 0.94 g/cm³
Flash Pt: 93 °C
MDL Number: MFCD00000772
CAS Number: 38460-95-6
EINECS: 253-951-0
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4532 to 1.4552 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty