Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trifluoromethanesulphonyl chloride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17305-0050 17305-0250 17305-1000
TS17305-0050EA 76 USD InStock
TS17305-0050 TS17305-0250 TS17305-1000
Trifluoromethanesulphonyl chloride 99%
Trifluoromethanesulphonyl chloride


Formula: CClF₃O₂S
MW: 168.52
Boiling Pt: 32
Density: 1.58 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00007451
CAS Number: 421-83-0
EINECS: 207-009-0
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.3330 to 1.3350 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty