Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-butene 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17355-0050 17355-0250 17355-0500
TS17355-0050EA 114 USD InStock
TS17355-0050 TS17355-0250 TS17355-0500
4-Bromo-1-butene 98+%
4-Bromo-1-butene


Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 98
Density: 1.33 g/cm³
Flash Pt: 1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000258
CAS Number: 5162-44-7
EINECS: 225-937-4
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4595 to 1.4635 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty