Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethylacryloyl chloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17973-0050 17973-0250
TS17973-0050EA 60 USD InStock
TS17973-0050 TS17973-0250
3,3-Dimethylacryloyl chloride 97%
3,3-Dimethylacryloyl chloride


Formula: C₅H₇ClO
Boiling Pt: 145…147 °C (760 mmHg)
Density: 1.06 g/cm³
Flash Pt: 51 °C
Storage Temperature: Refrigerator
CAS Number: 3350-78-5
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4780 (20°C, 589 nm)
GC ≥96.0 %
Titration with NaOH 98.0 to 100.5 % (dried substance)
Chloride (Cl) ≤0.04 %
Sulfate (SO4) ≤0.1 %
Arsenic (As) ≤4 ppm
Iron (Fe) ≤40 ppm
Heavy metals (as Pb) ≤20 ppm
Impurity ≤2.5 % (Monosodium phosphate)
Insoluble matter ≤0.4 %
Reducing substances Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty