Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodobenzenesulphonyl chloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

20729-0050 20729-0250
TS20729-0050EA 52 USD InStock
TS20729-0050 TS20729-0250
4-Iodobenzenesulphonyl chloride 97%
4-Iodobenzenesulphonyl chloride


Formula: C₆H₄ClIO₂S
Melting Pt: 80…84 °C
MDL Number: MFCD00007441
CAS Number: 98-61-3
EINECS: 202-686-9
UN: 1759
ADR: 8,II
Order Now
Specification Test Results
Appearance White to beige to orange Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 80 to 84 °C
Titration Argentometric ≥96.0 % (ex Cl)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty