Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Bromoethyl)benzene 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

21839-0250 21839-1000
TS21839-0250EA 66 USD InStock
TS21839-0250 TS21839-1000
(1-Bromoethyl)benzene 97%
(1-Bromoethyl)benzene


Formula: C₆H₅CHBrCH₃
MW: 185.06 g/mol
Boiling Pt: 203
Melting Pt: 50
Density: 1.35 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00000139
CAS Number: 585-71-7
EINECS: 209-560-2
UN: 3334
ADR: 9 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f329f7855ae4dffcd097f49d6d504b1ec165e692&vid=f329f7855ae4dffcd097f49d6d504b1ec165e692&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5630 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty