Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-1-pentyne 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

22342-0250 22342-0050
TS22342-0250EA 166 USD InStock
TS22342-0250 TS22342-0050
5-Chloro-1-pentyne 98%
5-Chloro-1-pentyne


Formula: C₅H₇Cl
MW: 102.56 g/mol
Boiling Pt: 67
Density: 0.96 g/cm³
Flash Pt: 15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001014
CAS Number: 14267-92-6
EINECS: 238-159-5
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty