Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethyl-1-butyne 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

22344-0050 22344-0250
TS22344-0050EA 67 USD InStock
TS22344-0050 TS22344-0250
3,3-Dimethyl-1-butyne 98%
3,3-Dimethyl-1-butyne


Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 38
Melting Pt: -78.2
Density: 0.66 g/cm³
Flash Pt: –034 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008852
CAS Number: 917-92-0
EINECS: 213-035-3
UN: 3295
ADR: 3 I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3730 to 1.3750 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty