Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2-cyclopenten-1-one 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

27036-0010 27036-0050
TS27036-0010EA 71 USD InStock
TS27036-0010 TS27036-0050
2-Methyl-2-cyclopenten-1-one 97%
2-Methyl-2-cyclopenten-1-one


Formula: C₆H₈O
Boiling Pt: 158…161 °C (760 mmHg)
Density: 0.979 g/cm³
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012275
CAS Number: 1120-73-6
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4800 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty