Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Anisoyl chloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

27611-0050 27611-0250
TS27611-0050EA 47 USD InStock
TS27611-0050 TS27611-0250
2-Anisoyl chloride 97%
2-Anisoyl chloride


Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 137
Density: 1.146 g/cm³
Flash Pt: 84 °C
MDL Number: MFCD00000664
CAS Number: 21615-34-9
EINECS: 244-477-5
UN: 1729
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5714 to 1.5734 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty