Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Geranyl linalool mixture of isomers ∼95% tech.

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

27836-0025 27836-0250
TS27836-0025EA 62 USD InStock
TS27836-0025 TS27836-0250
Geranyl linalool mixture of isomers ∼95% tech.
Geranyl linalool


Formula: C₂₀H₃₄O
MW: 290.491 g/mol
Boiling Pt: 250 °C (760 mmHg)
Density: 0.88 g/cm³
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00059363
CAS Number: 1113-21-9
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4860 to 1.4920 (20°C, 589 nm)
GC ≥94 %
Specific gravity (25°C) 0.875 to 0.885

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty