Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Hydroxy-1-cyclopropanecarboxylic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30143-0010 30143-0050
TS30143-0010EA 144 USD InStock
TS30143-0010 TS30143-0050
1-Hydroxy-1-cyclopropanecarboxylic acid 97%
1-Hydroxy-1-cyclopropanecarboxylic acid


Formula: C₄H₆O₃
MW: 102.089 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010797
CAS Number: 17994-25-1
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty