Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecanedioyl dichloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30534-0050 30534-0250
TS30534-0050EA 65 USD InStock
TS30534-0050 TS30534-0250
Dodecanedioyl dichloride 98%
Dodecanedioyl dichloride


Formula: C₁₂H₂₀Cl₂O₂
MW: 267.19 g/mol
Boiling Pt: 140 °C (.5 mmHg)
Density: 1.06 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00012243
CAS Number: 4834-98-4
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4690 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty