Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Bis(methylthio)-2-nitroethylene 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

30569-0050 30569-0250
TS30569-0050EA 55 USD InStock
TS30569-0050 TS30569-0250
1,1-Bis(methylthio)-2-nitroethylene 99%
1,1-Bis(methylthio)-2-nitroethylene


Formula: C₄H₇NO₂S₂
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00010443
CAS Number: 13623-94-4
EINECS: 237-108-4
Order Now
Specification Test Results
Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 124 to 127 °C
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty