Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6-bromohexanoate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

30592-0250 30592-1000
TS30592-0250EA 71 USD InStock
TS30592-0250 TS30592-1000
Ethyl-6-bromohexanoate 98%
Ethyl-6-bromohexanoate


Formula: BrH₂C(CH₂)₄COOCH₂CH₃
Boiling Pt: 128…130 °C (16 mmHg)
Melting Pt: 33 °C
Density: 1.254 g/cm³
Flash Pt: 58,3 °C
MDL Number: MFCD00000270
CAS Number: 25542-62-5
EINECS: 247-085-2
UN: 3272
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4600 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty