Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)benzoyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30635-0010 30635-0050
TS30635-0010EA 37 USD InStock
TS30635-0010 TS30635-0050
4-(Trifluoromethoxy)benzoyl chloride 98%
4-(Trifluoromethoxy)benzoyl chloride


Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
Boiling Pt: 55…56 °C (2 mmHg)
Density: 1.42 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00052329
CAS Number: 36823-88-8
UN: 2920
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to pink Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4730 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty