Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nonafluorovaleric acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30929-0250 30929-0050
TS30929-0250EA 382 USD InStock
TS30929-0250 TS30929-0050
Nonafluorovaleric acid 97%
Nonafluorovaleric acid


Formula: C₅HF₉O₂
MW: 264.05 g/mol
Boiling Pt: 140 °C
Density: 1.71 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00040211
CAS Number: 2706-90-3
UN: 3265
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear light yellow to light brown Liquid
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty