Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetylenedicarboxylic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10252-0250 10252-1000
TS10252-0250EA 105 USD InStock
TS10252-0250 TS10252-1000
Acetylenedicarboxylic acid 98%
Acetylenedicarboxylic acid


Formula: C₄H₂O₄
Melting Pt: 180…187 °C (dec.)
Flash Pt: 131 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004362
CAS Number: 142-45-0
EINECS: 205-536-0
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to cream to beige Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty