Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetylindole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

10259-0050 10259-0250
TS10259-0050EA 62 USD InStock
TS10259-0050 TS10259-0250
3-Acetylindole 97%
3-Acetylindole


Formula: C₁₀H₉NO
Boiling Pt: 144…145 °C (10 mmHg)
Melting Pt: 185…191 °C
MDL Number: MFCD00005626
CAS Number: 703-80-0
EINECS: 211-875-5
Order Now
Specification Test Results
Appearance Yellow to light brown Powder
Infrared spectrum Conforms
Melting point 185 to 191 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty