Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylthiophene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10272-1000 10272-5000
TS10272-1000EA 53 USD InStock
TS10272-1000 TS10272-5000
2-Acetylthiophene 98%
2-Acetylthiophene


Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 214
Melting Pt: 9
Density: 1.16 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00005442
CAS Number: 88-15-3
EINECS: 201-804-6
UN: 2810
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Clear light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5640 to 1.5660 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty