Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminobutyraldehyde diethyl acetal 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10326-0050 10326-0250 10326-1000
TS10326-0010EA 22.66 USD InStock
TS10326-0010 TS10326-0050 TS10326-0250 TS10326-1000
4-Aminobutyraldehyde diethyl acetal 95%
4-Aminobutyraldehyde diethyl acetal


Formula: C₈H₁₉NO₂
Boiling Pt: 196 °C (760 mmHg)
Density: 0.9 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00008227
CAS Number: 6346-09-4
EINECS: 228-753-2
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4310 (20 °C, 589 nm)
GC ≥94 %
Solubility (10% in H2O) Clear colorless solution
Titration with HClO4 99.0 to 101.0 % (On dry substance)
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Heavy metals (as Pb) ≤10 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty