Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzoylpyridine 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10580-0250 10580-1000
TS10580-0250EA 33 USD InStock
TS10580-0250 TS10580-1000
2-Benzoylpyridine 99+%
2-Benzoylpyridine


Formula: C₁₂H₉NO
MW: 183.21 g/mol
Boiling Pt: 317 °C (1013 hPa)
Melting Pt: 42…44 °C
Flash Pt: 150 °C
MDL Number: MFCD00006300
CAS Number: 91-02-1
EINECS: 202-034-3
Order Now
Specification Test Results
Appearance Colorless to light yellow Crystalline chunks
Infrared spectrum Conforms
Melting point 42 to 44 °C
GC ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty