Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-tert-Butylcyclohexanol mixture of isomers 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10790-0050 10790-2500
TS10790-0050EA 29 USD InStock
TS10790-0050 TS10790-2500
4-tert-Butylcyclohexanol mixture of isomers 99%
4-tert-Butylcyclohexanol


Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 214
Melting Pt: 57
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00001473
CAS Number: 98-52-2
EINECS: 202-676-4
Order Now
Specification Test Results
Appearance White Powder or granules
Infrared spectrum Conforms
Melting point 60 to 69 °C (depending on cis/trans ratio)
GC ≥98.5 %
Ratio Cis/trans ratio: approximately 70% trans, 30% cis
Specific optical rotation -12° to -18° (20 °C, 589 nm) (neat)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty