Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl dichlorophosphate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11838-0050 11838-0250 11838-1000
TS11838-0050EA 37 USD InStock
TS11838-0050 TS11838-0250 TS11838-1000
Ethyl dichlorophosphate 97%
Ethyl dichlorophosphate


Formula: C₂H₅Cl₂O₂P
Boiling Pt: 167 °C (758 mmHg)
Density: 1.373 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002069
CAS Number: 1498-51-7
EINECS: 216-099-0
UN: 3390
ADR: 6.1,
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20 °C, 589 nm)
GC ≥96.0 %
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤30 ppm
Lead (Pb) ≤100 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty