Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Hydroxyacetophenone 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12093-1000 12093-5000
TS12093-1000EA 32 USD InStock
TS12093-1000 TS12093-5000
2'-Hydroxyacetophenone 99%
2'-Hydroxyacetophenone


Formula: C₈H₈O₂
Boiling Pt: 92…93 °C (12 mmHg)
Melting Pt: 4…6 °C
Density: 1.131 g/cm³
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00002219
CAS Number: 118-93-4
EINECS: 204-288-0
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5580 to 1.5600 (20 °C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty