Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Hydroxy-2'-methylacetophenone 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12139-0050 12139-0250 12139-1000
TS12139-0050EA 52 USD InStock
TS12139-0050 TS12139-0250 TS12139-1000
4'-Hydroxy-2'-methylacetophenone 97%
4'-Hydroxy-2'-methylacetophenone


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 313 °C (760 mmHg)
Melting Pt: 127…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00002303
CAS Number: 875-59-2
EINECS: 212-874-2
Order Now
Specification Test Results
Appearance Off-white to beige to tan Powder or crystals
Infrared spectrum Conforms
Melting point 127 to 132 °C
HPLC ≥96.0 %
Color scale ≤20 APHA
Acidity/alkalinity ≤0.00008 meq/g
Substance darkened by H2SO4 passes test
Calcium (Ca) ≤0.2 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Hydrocarbons ≤0.1 % Aromatic hydrocarbons
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤10 ppm
Bromine number ≤0.5

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty