Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Malonyl dichloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

12528-0250 12528-1000
TS12528-0250EA 134 USD InStock
TS12528-0250 TS12528-1000
Malonyl dichloride 97%
Malonyl dichloride


Formula: C₃H₂Cl₂O₂
MW: 140.95 g/mol
Boiling Pt: 47 °C (15 mmHg)
Density: 1.45 g/cm³
Flash Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000735
CAS Number: 1663-67-8
EINECS: 216-772-9
UN: 2920
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear yellow to orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4660 (20 °C, 589 nm)
Titration Argentometric ≥96.0 %
Residue after evaporation ≤0.0003 %
Conductivity ≤1 µS/cm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty