Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Methylacetophenone 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12618-0010 12618-0050 12618-2500
TS12618-0010EA 99 USD InStock
TS12618-0010 TS12618-0050 TS12618-2500
4'-Methylacetophenone 95%
4'-Methylacetophenone


Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 226
Melting Pt: 28
Density: 1.005 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008751
CAS Number: 122-00-9
EINECS: 204-514-8
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5350 (20 °C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty