Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methylcyclohexanol 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12660-0050 12660-0250 12660-1000
TS12660-0050EA 37 USD InStock
TS12660-0050 TS12660-0250 TS12660-1000
1-Methylcyclohexanol 96%
1-Methylcyclohexanol


Formula: C₇H₁₄O
Boiling Pt: 168 °C (752 mmHg)
Melting Pt: 26 °C
Density: 0.919 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00003857
CAS Number: 590-67-0
EINECS: 215-963-4
UN: 1325
ADR: 4.1,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20 °C, 589 nm)
GC ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty