Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Naphthaldehyde 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12810-0050 12810-0250 12810-1000
TS12810-0050EA 54 USD InStock
TS12810-0050 TS12810-0250 TS12810-1000
2-Naphthaldehyde 98%
2-Naphthaldehyde


Formula: C₁₁H₈O
Boiling Pt: 160 °C (19 mmHg)
Melting Pt: 58…62 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004094
CAS Number: 66-99-9
EINECS: 200-640-2
UN: 3335
ADR: 9,III
Order Now
Specification Test Results
Appearance Off-white to yellow to light beige Powder and/or chunks
Infrared spectrum Conforms
Melting point 58 to 62 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty