Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Nitrobenzyl)pyridine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12855-0050 12855-0100 12855-0500
TS12855-0050EA 50 USD InStock
TS12855-0050 TS12855-0100 TS12855-0500
4-(4-Nitrobenzyl)pyridine 98%
4-(4-Nitrobenzyl)pyridine


Formula: C₁₂H₁₀N₂O₂
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006445
CAS Number: 1083-48-3
EINECS: 214-108-2
Order Now
Specification Test Results
Appearance Light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 69 to 72 °C
Titration with HClO4 ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty