Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pimelic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

13123-0250 13123-1000 13123-5000
TS13123-0250EA 77 USD InStock
TS13123-0250 TS13123-1000 TS13123-5000
Pimelic acid 98%
Pimelic acid


Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 354
Melting Pt: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00004425
CAS Number: 111-16-0
EINECS: 203-840-8
Merck Index: 12,07585
Order Now
Specification Test Results
Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 103 to 106 °C
Titration with NaOH 97.5 to 102.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty