Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobutyric acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

13347-0050 13347-0250 13347-1000
TS13347-0050EA 41 USD InStock
TS13347-0050 TS13347-0250 TS13347-1000
4-Bromobutyric acid 98%
4-Bromobutyric acid


Formula: BrH₂C(CH₂)₂COOH
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 28…32 °C
Flash Pt: 124 °C
MDL Number: MFCD00002817
CAS Number: 2623-87-2
EINECS: 220-083-9
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Appearance Light brown Low melting crystals
Infrared spectrum Conforms
Melting point 28 to 32 °C
Titration with NaOH 97.5 to 102.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty