Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Heptanone 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

14655-0025 14655-0050 14655-5000
TS14655-0025EA 412 USD InStock
TS14655-0025 TS14655-0050 TS14655-5000
4-Heptanone 98%
4-Heptanone


Formula: C₇H₁₄O
Boiling Pt: 145 °C (760 mmHg)
Melting Pt: –33 °C
Density: 0.81 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00009403
CAS Number: 123-19-3
EINECS: 204-608-9
UN: 2710
ADR: 3,III
Merck Index: 13,03379
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4080 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty