Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

14910-0050 14910-0250 14910-1000
TS14910-0050EA 30 USD InStock
TS14910-0050 TS14910-0250 TS14910-1000
5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%
5-Norbornene-2-carboxaldehyde


Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 67…70 °C (12 mmHg)
Density: 1.018 g/cm³
Flash Pt: 50.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00167568
CAS Number: 5453-80-5
UN: 1989
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4920 (20 °C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty