Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxypropiononitrile 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

14939-0010 14939-0500 14939-2500
TS14939-0010EA 274 USD InStock
TS14939-0010 TS14939-0500 TS14939-2500
3-Hydroxypropiononitrile 99%
3-Hydroxypropiononitrile


Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 228
Melting Pt: -46
Density: 1.04 g/cm³
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002826
CAS Number: 109-78-4
EINECS: 203-704-8
Merck Index: 13,03828
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4246 to 1.4266 (20 °C, 589 nm)
GC ≥98.5 %
Specific optical rotation -28° to -32° (20°C, 589 nm) (c=1 in DMF)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty