Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl phenylmalonate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15026-0010 15026-0025 15026-1000 15026-2500
TS15026-0010EA 128 USD InStock
TS15026-0010 TS15026-0025 TS15026-1000 TS15026-2500
Diethyl phenylmalonate 98%
Diethyl phenylmalonate


Formula: C₁₃H₁₆O₄
MW: 236.27 g/mol
Boiling Pt: 170…172 °C (14 mmHg)
Melting Pt: 16…17 °C
Density: 1.09 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009144
CAS Number: 83-13-6
EINECS: 201-456-5
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4903 to 1.4923 (20 °C, 589 nm)
Water ≤0.05 %
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty