Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromo-4-methoxyacetophenone 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15190-0050 15190-0250 15190-1000
TS15190-0050EA 41 USD InStock
TS15190-0050 TS15190-0250 TS15190-1000
ω-Bromo-4-methoxyacetophenone 98%
ω-Bromo-4-methoxyacetophenone


Formula: C₉H₉BrO₂
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 69…72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000201
CAS Number: 2632-13-5
EINECS: 220-118-8
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Off-white to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 69 to 72 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty