Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl methylmalonate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15514-1000 15514-5000
TS15514-1000EA 55 USD InStock
TS15514-1000 TS15514-5000
Diethyl methylmalonate 99%
Diethyl methylmalonate


Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Density: 1.01 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009162
CAS Number: 609-08-5
EINECS: 210-175-7
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20 °C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty