Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethyl-2-butanol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15658-0050 15658-0250
TS15658-0050EA 80 USD InStock
TS15658-0050 TS15658-0250
2,3-Dimethyl-2-butanol 98%
2,3-Dimethyl-2-butanol


Formula: C₆H₁₄O
MW: 102.18
Boiling Pt: 119
Melting Pt: -10.4
Density: 0.82 g/cm³
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00004463
CAS Number: 594-60-5
EINECS: 209-847-2
UN: 1987
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty