Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-2-pyrrolecarboxaldehyde 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15693-0050 15693-0250
TS15693-0050EA 40 USD InStock
TS15693-0050 TS15693-0250
1-Methyl-2-pyrrolecarboxaldehyde 98%
1-Methyl-2-pyrrolecarboxaldehyde


Formula: C₆H₇NO
Boiling Pt: 87…90 °C (22 mmHg)
Density: 1.01 g/cm³
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003087
CAS Number: 1192-58-1
Order Now
Specification Test Results
Appearance Clear colorless to orange-red to pink-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5580 to 1.5630 (20 °C, 589 nm)
GC ≥97.5 %
Carbonate (CO3) Passes test
Sulfate (SO4) ≤0.5 %
Arsenic (As) ≤0.0003 %
Barium (Ba) Passes test
Iron (Fe) ≤0.04 %
Insoluble matter ≤0.2 % (in HCl)
Loss on ignition 6.6 % to 8.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty