Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Methyl-1-butanol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15827-0010 15827-0025 15827-0250 15827-1000
TS15827-0010EA 70 USD InStock
TS15827-0010 TS15827-0025 TS15827-0250 TS15827-1000
(±)-2-Methyl-1-butanol 98%
(±)-2-Methyl-1-butanol


Formula: C₅H₁₂O
Boiling Pt: 129 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.81 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00004743
CAS Number: 137-32-6
EINECS: 205-289-9
UN: 1105
ADR: 3,III
Merck Index: 12,06109
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4120 (20 °C, 589 nm)
GC ≥97.5 %
Specific optical rotation -0.5° to +0.5° (20°C, 589 nm) (Neat)
Sulfate (SO4) ≤0.05 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty