Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethyl-3-pentanone 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15970-2500 15970-5000
TS15970-2500EA 77 USD InStock
TS15970-2500 TS15970-5000
2,4-Dimethyl-3-pentanone 98%
2,4-Dimethyl-3-pentanone


Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 124
Melting Pt: -69
Density: 0.8 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00008918
CAS Number: 565-80-0
EINECS: 209-294-7
UN: 1224
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4020 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty